Order Entry
Austria
ContactUsLinkComponent
1,4-Dioxane
Flammable
Health
Irritant
1,4-Dioxane
Catalog # EHERC12865000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Flammable
Health
Irritant
1,4-Dioxane
Catalog # EHERC12865000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :88.11 g/mol
Merck Index:13,03330
Boiling Pt:101.1 °C (1013 hPa)
Melting Pt:11.8 °C
Density:1.03 g/cm³ (20 °C)
Flash Pt:12 °C
EINECS:204-661-8
UN:1165
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:123-91-1
Formula:C₄H₈O₂
MDL Number:MFCD00006571