


1,4-Dioxane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Merck Index:13,03330
Boiling Pt:101 °C (760 mmHg)
Melting Pt:12 °C
Density:1.034 g/cm³
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:204-661-8
UN:1165
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.03 %
- GC:≥99.0 %
- UV:at 220 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
Specification Test Results
Frequently Bought Together