Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Methylcyclohexane ≥99%

Flammable
Health
Irritant
Environment
Methylcyclohexane ≥99%
Certificates
Methylcyclohexane ≥99%
Methylcyclohexane ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 108-87-2 Molecular Formula: C7H14 Formula Weight: 98.19 Flash Point: -4°(-25°F) Physical Form: Liquid Density: 0.771 Refractive Index: 1.4230 Appearance: Colourless Melting Point: -126° Boiling Point: 101° Beilstein Reference: 505972 Merck Reference: 14,6047 UN No.: 2296 MDL No.: MFCD00001497

  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:14,6047
  • Boiling Pt:101 °C
  • Melting Pt:–126 °C
  • Density:0.771
  • Flash Pt:–4 °C (–25 °F)
  • Storage Temperature:Ambient
  • EINECS:203-624-3
  • UN:2296
  • ADR:3,II
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.