2-Methylcyclohexanone
Catalog # EHERC15085010
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:112.17 g/mol
Boiling Pt:163 °C (1013 hPa)
Melting Pt:–14 °C
Density:0,924 g/cm³ (20 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:209-513-6
UN:2297
ADR:3,III
:GHS02
:GHS07
CAS Number:583-60-8
Formula:C₇H₁₂O
MDL Number:MFCD00001635