Order Entry
Austria
Orders LinkContactUsLinkComponent
Cyclopentane ≥97%, TECHNICAL

Cyclopentane ≥97%, TECHNICAL

Glass bottle, 100 mL
Catalog # 23251.183
Flammable
Supplier:   VWR Chemicals
Cyclopentane ≥97%, TECHNICAL
Catalog # 23251.183
Supplier:  VWR Chemicals
CAS Number:  287-92-3
Cyclopentane ≥97%, TECHNICAL
Cyclopentane ≥97%, TECHNICAL

Glass bottle, 100 mL

CAS Number:  287-92-3
Price
126,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Pentamethylene
  • CAS Number:287-92-3
  • Formula:C₅H₁₀
  • MDL Number:MFCD00001356
  • Molecular Weight:70.13 g/mol
  • Merck Index:13,02769
  • Boiling Pt:49 °C (1013 hPa)
  • Melting Pt:–93,3 °C
  • Density:0.746 g/cm³ (20 °C)
  • Flash Pt:–41 °C
  • Storage Temperature:Ambient
  • EINECS:206-016-6
  • UN:1146
  • ADR:3,II
Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • IR Spectrum:
    Passes test
  • Density (20/4):
    0.740 - 0.745

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications