Brij® 35 30% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
CAS Number:9002-92-0
:
Formula:C₅₈H₁₁₈O₂₄
MDL Number:MFCD00080891
:1199.56 g/mol
:M-Clarity™ quality level = MQ200
EINECS:500-002-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together