Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Hexahydrobenzene, Hexamethylene
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–18 °C
REACH:01-2119463273-41
:GHS02
:GHS08
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.5 %
- Evaporation residue:Max. 5 ppm
- Water:Max. 0.02 %
- Pesticide analysis (Lindane/ECD):Max. 5 ng/l
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
Frequently Bought Together