3-Bromo-5-hydroxybenzoic acid, Sigma-Aldrich®
Catalog # SIAL686417-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:217.02 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:140472-69-1
Formula:C₇H₅BrO₃
MDL Number:MFCD06797980