Saccharin sodium
Catalog # PROOMM0818.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1,2-Benzothiazol-3(2H)-one 1,1-dioxide sodium salt
:
:206.18 g/mol
EINECS:204-886-1
CAS Number:128-44-9
Formula:C₇H₅NNaO₃S
MDL Number:MFCD00013092