TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Catalog # MOLE21958982-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid
:
:243.28 g/mol
Storage Temperature:Ambient
EINECS:249-954-1
CAS Number:29915-38-6
Formula:C₇H₁₇NO₆S
MDL Number:MFCD00007538