Dibutyltin dilaurate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Di-n-butyltin dilaurate
CAS Number:77-58-7
:
Formula:C₃₂H₆₄O₄Sn
MDL Number:MFCD00008963
:631.57 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:201-039-8
UN:3261
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:- 0.1 g/l (20 °C)
- Melting Point:25 - 27 °C
- Molar Mass:631.55 g/mol
- Boiling Point:250 °C (1013 hPa)
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:149 °C
- Density:1.05 g/cm3 (20 °C)
- Ignition Point:200 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Related Information
Frequently Bought Together