



Cyclohexane, Uvasol® for spectroscopy, Supelco®
: Merck
:110-82-7
:
:C₆H₁₂
:MFCD00003814
:84.16 g/mol
:13,02752
:81 °C (1013 hPa)
:7 °C
:0.7785 g/cm³ (20 °C)
:Ambient
:203-806-2
:1145
:3,II
:M-Clarity™ quality level = MQ100
:–18 °C
:GHS02
:GHS08
:GHS07
:GHS09
- Solubility:55 mg/L (20 °C)
- Melting Point:6 °C
- Molar Mass:84.16 g/mol
- Boiling Point:81 °C (1013 hPa)
- Vapor Pressure:103 hPa (20 °C)
- Flash Point:-18 °C
- Refractive Index:1.4264 (20 °C)
- Explosion Limit:1.2 - 8.3%(V)
- Density:0.78 g/cm³ (20 °C)
- pH:(H₂O) not applicable
- Ignition Point:260 °C
- Viscosity Kinematic:1.26 mm²/s (20 °C)
- Evaporation Number:3.5
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together