
Brij® 35 10% in aqueous solution, Proteomics Grade
Supplier: G-Biosciences
Certificates
About this item
CAS Number:9002-92-0
:
Formula:C₅₈H₁₁₈O₂₄
MDL Number:MFCD00080891
:1199.56 g/mol
EINECS:500-002-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together