TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Catalog # 786-513
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
:
Merck Index:13,06793
Boiling Pt:∼200 °C (1013 hPa)
Melting Pt:6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:251 °C (closed cup)
UN:3082
ADR:9,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Refrigerator
CAS Number:9002-93-1
:9036-19-5
MDL Number:MFCD00132505