Bis(2-ethylhexyl) adipate
Catalog # EHERC10046000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:370.57 g/mol
EINECS:203-090-1
:GHS07
CAS Number:103-23-1
Formula:C₂₂H₄₂O₄
MDL Number:MFCD00009496