Order Entry
Austria
ContactUsLinkComponent
Vitamin A acetate
Vitamin A acetate
Catalog # EDQMR0300000
Supplier:  EDQM
CAS Number:  
Vitamin A acetate
Catalog # EDQMR0300000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :328.49 g/mol
EINECS:204-844-2
CAS Number:127-47-9
Formula:C₂₂H₃₂O₂
MDL Number:MFCD00019413