Order Entry
Austria
ContactUsLinkComponent
Brij® 35 10% in aqueous solution, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®
Brij® 35 10% in aqueous solution, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®
Catalog # 203728-50
CAS Number:  
Brij® 35 10% in aqueous solution, PROTEIN GRADE® detergent, sterile-filtered, Calbiochem®, Millipore®
Catalog # 203728-50
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :1199.56 g/mol
EINECS:500-002-6
 :GHS07
Storage Temperature:Refrigerator
CAS Number:9002-92-0
Formula:C₅₈H₁₁₈O₂₄
MDL Number:MFCD00080891