Brij® 35 30% in aqueous solution non-ionic detergent, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Synonyms: Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
CAS Number:9002-92-0
:
Formula:C₅₈H₁₁₈O₂₄
MDL Number:MFCD00080891
:1199.56 g/mol
EINECS:500-002-6
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together