


Lithium bis(trifluoromethylsulphonyl)imide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:90076-65-6
:
Formula:C₂F₆LiNO₄S₂
MDL Number:MFCD00210017
Melting Pt:234…238 °C
Storage Temperature:Ambient
EINECS:415-300-0
UN:2923
ADR:8,II
:287.09 g/mol
:GHS06
:GHS05
:GHS08
Related Information
Frequently Bought Together