

Ethyl vinyl ether 99% stabilised
:109-92-2
:
:C₄H₈O
:MFCD00009248
:72.11 g/mol
:36 °C (760 mmHg)
:–116 °C
:0.75 g/cm³
:–45 °C
:1302
:3,I
:Stabilised with potassium hydroxide
:203-718-4
:GHS02
:GHS07
:Refrigerator
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3760 to 1.3780 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:0.1 % KOH
Frequently Bought Together