Order Entry
Austria
Orders LinkContactUsLinkComponent
Quinoline ≥96%, TECHNICAL

Quinoline ≥96%, TECHNICAL

Glass bottle, 1 L
Catalog # 27341.294
Health
Irritant
Environment
Supplier:   VWR Chemicals
Quinoline ≥96%, TECHNICAL
Catalog # 27341.294
Supplier:  VWR Chemicals
CAS Number:  91-22-5
Quinoline ≥96%, TECHNICAL
Quinoline ≥96%, TECHNICAL

Glass bottle, 1 L

CAS Number:  91-22-5
Price
190,00 €
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:91-22-5
  • Formula:C₉H₇N
  • MDL Number:MFCD00006736
  • Molecular Weight:129.16 g/mol
  • Merck Index:13,08160
  • Boiling Pt:238 °C (1013 hPa)
  • Melting Pt:–15 °C
  • Density:1.095 g/cm³ (20 °C)
  • Flash Pt:107 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:202-051-6
  • UN:2656
  • ADR:6.1,III
Specifications

Specification Test Results

  • Assay:
    Min. 96 %
  • IR Spectrum:
    Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.