Order Entry
Austria
Orders LinkContactUsLinkComponent
Mesitylene for synthesis

Mesitylene for synthesis

Flammable
Health
Irritant
Environment
Supplier:   Merck
Mesitylene for synthesis
Supplier:  Merck
Certificates
Mesitylene for synthesis

2 Options Available Below

Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:165 °C (1013 hPa)
  • Melting Pt:–45 °C
  • Density:0.8637 g/cm³ (20 °C)
  • Flash Pt:48 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.