Order Entry
Austria
Orders LinkContactUsLinkComponent
Mesitylene ≥98%

Mesitylene ≥98%

Flammable
Irritant
Environment
Mesitylene ≥98%
Certificates
Mesitylene ≥98%
Mesitylene ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 108-67-8 Molecular Formula: C9H12 Formula Weight: 120.20 Flash Point: 44°(111°F) Refractive Index: 1.4990 Odour: Unpleasant Melting Point: -45° Boiling Point: 163-165° Beilstein Reference: 906806 Merck Reference: 14,5907 UN No.: 2325 MDL No.: MFCD00008538

Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:14,5907
  • Boiling Pt:163…165 °C
  • Melting Pt:–45 °C
  • Density:0.865
  • Flash Pt:44 °C (111 °F)
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.