Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Mesitylene 99%, extra pure

Irritant
Environment
Flammable
Health
Mesitylene 99%, extra pure
Certificates
Mesitylene 99%, extra pure

3 Options Available Below

Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:163…166 °C (760 mmHg)
  • Melting Pt:–45 °C
  • Density:0.8675 g/cm³
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4980 to 1.5000 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Residue after evaporation:
    ≤0.01 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 56,40 €
    56,40 €
    1 * 250 mL
       56,40 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 139,00 €
    139,00 €
    1 * 1 L
       139,00 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 248,00 €
    248,00 €
    1 * 2,5 L
       248,00 €