
1,3,5-Triethylbenzene
Catalog # EHERC17833500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:162.27 g/mol
Boiling Pt:215 °C (1013 hPa)
Boiling Pt:215 °C (1013 hPa)
Melting Pt:3 °C
Density:0.8631 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-017-3
UN:3082
ADR:9,III
:GHS07
CAS Number:102-25-0
Formula:C₁₂H₁₈
MDL Number:MFCD00009261