Order Entry
Austria
Orders LinkContactUsLinkComponent
2,4,5-Trimethylphenol ≥99%

2,4,5-Trimethylphenol ≥99%

2,4,5-Trimethylphenol ≥99%
Certificates
2,4,5-Trimethylphenol ≥99%
2,4,5-Trimethylphenol ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 496-78-6 Molecular Formula: C9H12O Formula Weight: 136.20 Odour: Phenol-like Melting Point: 72° Boiling Point: 232° UN No.: 2430 MDL No.: MFCD00020050

  • CAS Number:496-78-6
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00020050
  • Molecular Weight:136.19 g/mol
  • Boiling Pt:232 °C
  • Melting Pt:72 °C
  • Storage Temperature:Ambient
  • EINECS:207-832-5
  • UN:2430
  • ADR:8,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.