Orders LinkContactUsLinkComponent
Watchmaker forceps
Supplier:   MURRAY S
Watchmaker forceps
Supplier:  MURRAY S
Certificates
Watchmaker forceps
Watchmaker forceps
4 Options Available Below
Product Details & Documents
Stainless steel. Ideal for small-scale dissection applications or precise part placement.
  • Very fine points
  • Smooth flat handle
  • Available straight or curved
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Watchmaker No. 7, curved
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,10 €
    12,10 €
    1 * 1 ST
       12,10 €
  • Watchmaker No. 4, straight
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,10 €
    12,10 €
    1 * 1 ST
       12,10 €
  • Watchmaker No. 3, straight
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,10 €
    12,10 €
    1 * 1 ST
       12,10 €
  • Watchmaker No. 5, straight
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,10 €
    12,10 €
    1 * 1 ST
       12,10 €