2,6-Di-tert-butyl-p-cresol Ph. Eur., NF, E321, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:128-37-0
:
Formula:C₁₅H₂₄O
MDL Number:MFCD00011644
:220.35 g/mol
Merck Index:13,01547
Boiling Pt:265 °C (1013 hPa)
Melting Pt:69…70 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:127 °C
Storage Temperature:Ambient
EINECS:204-881-4
UN:3077
ADR:9,III
:GHS09
Related Information
Frequently Bought Together