2,6-Di-tert-butyl-p-cresol 99.8%
:128-37-0
:
:C₁₅H₂₄O
:MFCD00011644
:220.35 g/mol
:13,01547
:265 °C (760 mmHg)
:69…71 °C
:1,05 g/cm³ (20 °C)
:127 °C
:Ambient
:204-881-4
:3077
:9,III
:GHS09
- Appearance:Semitransparent white Crystals
- Infrared spectrum:Conforms
- Melting point:69 to 71 °C
- Water:≤0.12 % (K.F.)
- GC:≥99.8 %
- Color of solution:≤30 APHA (30 % w/w in acetone)
Frequently Bought Together