p-Ethyltoluene, Sigma-Aldrich®
Catalog # SIAL04943-10ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:120.19 g/mol
Boiling Pt:162 °C (1013 hPa)
Melting Pt:–62 °C
Density:0.8614 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-761-2
UN:3295
ADR:3,III
:M-Clarity™ quality level = MQ100
CAS Number:622-96-8
Formula:C₉H₁₂
MDL Number:MFCD00009263