ContactUsLinkComponent
Cytology funnels, MEGA CytoSep™, for SHANDON Cytospin® 4 cytocentrifuge
Cytology funnels, MEGA CytoSep™, for SHANDON Cytospin® 4 cytocentrifuge
Catalog # 720-1867
Supplier:  Simport Scientific
undefined
Cytology funnels, MEGA CytoSep™, for SHANDON Cytospin® 4 cytocentrifuge
Catalog # 720-1867
Supplier:  Simport Scientific
Specifications
  • For:
    Shandon Cytospin® 4 cytocentrifuge
  • Pk:
    480
Specifications
About this item
The CytoSep™ MEGA funnel provides an easy, efficient and cost effective method of producing high quality thin layer slide preparations. It eliminates the need for stainless steel slide clips, offering laboratory workflow improvements. It is dedicated for convenient preparation of larger volume samples. This larger funnel is quick and easy to remove. The large rectangular cell deposition area of 22×14,75 mm (325 mm²) provides for up to 12 times the sample volume (6 ml) of the single CytoSep™ cytology funnel. It produces more cost-effective thin layer preparations when compared to other thin layer methods. Strategically placed baffles inhibit cell settling which results in uniform cell deposition and excellent quality slide preparations. CytoSep™ cytology MEGA funnel and cap minimise user exposure to pathogens while reducing the risk of specimen cross-contamination. Can prepare both air-dried and fixed preparations.