Order Entry
Austria
ContactUsLinkComponent
 
Cover mat for deep 96-well plates
  RATI8101101
 :  RATIOLAB
undefined
Cover mat for deep 96-well plates
  RATI8101101
 :  RATIOLAB

 

  • Packed:
    cardboard box (10 × 10)
  • Description:
    Cover mat for 1,2 ml deep-96-well plate, low profile
  • Material:
    TPE
  • Pk:
    100

 

 

Closures for 96-Well Deep-Well-Plates Low-Profile, 1,2 ml. Made of thermo-plastic elastomer (TPE).

Autoclavable at 121 °C, 20 minutes.