Stirrer bearings, with cooling jacket, GL thread, KPG®
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
Stirrer bearings with NS cone and cooling water connections with thread size GL14.
- Surface ground and polished
Frequently Bought Together