Stirrer bearings, with vacuum jacket, KPG®
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
Stirrer bearings with NS cone and vacuum connections which made of precision calibrated glass tube.
- Surface is grounded and polished
- Interchangeable
Frequently Bought Together