ContactUsLinkComponent
Stirrer bearings, KPG®
Stirrer bearings, KPG®
Catalog # LENZ06300116
undefined
Stirrer bearings, KPG®
Catalog # LENZ06300116
Specifications
  • For:
    Shaft - Ø: 16 mm
  • Description:
    Stirrer bearing, HB 16, DURAN®
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
These stirrer bearings are made of precision calibrated glass tube.
  • Surface is grounded and polished
  • Interchangeable