ContactUsLinkComponent
Luminescence microplates, 96-well
Luminescence microplates, 96-well
Catalog # RATI6018710
Supplier:  RATIOLAB
undefined
Luminescence microplates, 96-well
Catalog # RATI6018710
Supplier:  RATIOLAB
Specifications
  • No. of wells:
    96
  • Sterile:
    +
  • Colour:
    Black
  • Pk:
    100
  • Well volume:
    300 μl
Specifications
About this item
These wells are with raised rims for reduced cross-contamination and effective sealing with cover films. Precise design, perfectly shaped for use with automatic systems.
  • SBS standard micro test plate format
  • 8×12 wells alphanumerically coded
  • Optimum space for precise positioning
  • Space-saving and stackable
  • Surface untreated