Order Entry
Austria
ContactUsLinkComponent
o-Xylene for synthesis, Sigma-Aldrich®
Flammable
Health
Irritant
o-Xylene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Health
Irritant
o-Xylene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    0.18 g/l (20 °C)
  • Melting Point:
    -25 °C
  • Molar Mass:
    106.17 g/mol
  • Boiling Point:
    144.4 °C (1013 hPa)
  • Vapor Pressure:
    7 hPa (20 °C)
  • Flash Point:
    30 °C
  • Explosion Limit:
    1.0 - 7.6 %(V)
  • Density:
    0.88 g/cm3 (20 °C)
  • Ignition Point:
    465 °C
  • Evaporation Number:
    13.5

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together