Orders LinkContactUsLinkComponent
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Supplier:   BURKLE
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Supplier:  BURKLE
Certificates
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
5 Options Available Below
Product Details & Documents
Borosilicate glass 3.3, class B.
  • Adjusted for Ex
  • Blue ring graduations, Schellbach stripe
  • Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet
Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Certifications: Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Product Family Options
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 5 ml
    0,05 ml
    0,015 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,00 €
    63,00 €
    1 * 1 ST
       63,00 €
  • 10 ml
    0,05 ml
    0,030 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,00 €
    63,00 €
    1 * 1 ST
       63,00 €
  • 15 ml
    0,1 ml
    0,045 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,00 €
    63,00 €
    1 * 1 ST
       63,00 €
  • 50 ml
    0,1 ml
    0,045 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,40 €
    73,40 €
    1 * 1 ST
       73,40 €
  • 25 ml
    0,1 ml
    0,045 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,00 €
    63,00 €
    1 * 1 ST
       63,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Spare nozzle, PP
    Spare nozzle, PP
    BURK9695-0040
    Log in for availability
    1 * 1 ST - 2,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Cap without feed tube for 1000 ml bottle
    Cap without feed tube for 1000 ml bottle
    BURK9695-0528
    Log in for availability
    1 * 1 ST - 1,60 €