Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
Supplier:   BURKLE
Measuring scoops
Supplier:  BURKLE
Certificates
Measuring scoops
Measuring scoops
8 Options Available Below
Product Details & Documents
Universal graduated PP shovels for filling and measuring of powders and granules. For exact metering, the graduated shovel is to be overfilled and the excess at the upper edge of the shovel is to be wiped off.
  • High chemical resistance
  • Calibrated volume when full to the brim
  • Suitable for food contact
Product Family Options
Product Information
  • Capacity
    Length
    Width
    Material
    Colour
    Availability
    Price
  • 5 ml
    82 mm
    27 mm
    PP
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,70 €
    0,70 €
    1 * 1 ST
       0,70 €
  • 10 ml
    100 mm
    34 mm
    PP
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,70 €
    0,70 €
    1 * 1 ST
       0,70 €
  • 25 ml
    135 mm
    44 mm
    PP
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,10 €
    1,10 €
    1 * 1 ST
       1,10 €
  • 50 ml
    160 mm
    55 mm
    PP
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,50 €
    1,50 €
    1 * 1 ST
       1,50 €
  • 100 ml
    200 mm
    70 mm
    PP
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,30 €
    2,30 €
    1 * 1 ST
       2,30 €
  • 250 ml
    260 mm
    90 mm
    PP
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,80 €
    4,80 €
    1 * 1 ST
       4,80 €
  • 500 ml
    315 mm
    118 mm
    PP
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,20 €
    7,20 €
    1 * 1 ST
       7,20 €
  • 1000 ml
    385 mm
    134 mm
    PP
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,40 €
    8,40 €
    1 * 1 ST
       8,40 €