Lidocaine, Sigma-Aldrich®
Catalog # SIALL1026-1VL
Supplier: Merck
CAS Number:
Specifications
- Pk:1 Vial
- Seq:0032
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
:234.34 g/mol
EINECS:205-302-8
CAS Number:137-58-6
Formula:C₁₄H₂₂N₂O
MDL Number:MFCD00026733