Order Entry
Austria
ContactUsLinkComponent
2,2-Diphenylethylamine, Sigma-Aldrich®
2,2-Diphenylethylamine, Sigma-Aldrich®
Catalog # SIALD206709-5G
Supplier:  Merck
CAS Number:  
2,2-Diphenylethylamine, Sigma-Aldrich®
Catalog # SIALD206709-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :197.28 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:223-565-7
CAS Number:3963-62-0
Formula:C₁₄H₁₅N
MDL Number:MFCD00008143