Order Entry
Austria
ContactUsLinkComponent
Metanil yellow, Sigma-Aldrich®
Metanil yellow, Sigma-Aldrich®
Catalog # SIAL64010-25G-F
Supplier:  Merck
CAS Number:  
Metanil yellow, Sigma-Aldrich®
Catalog # SIAL64010-25G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :375.38 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:209-608-2
CAS Number:587-98-4
Formula:C₁₈H₁₄N₃NaO₃S
MDL Number:MFCD00007486