Zinc chloride 0.1 mol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
:136.3 g/mol
Storage Temperature:Refrigerator
EINECS:231-592-0
UN:3082
ADR:9,III
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together