Order Entry
Austria
ContactUsLinkComponent
Lidocaine
Lidocaine
Catalog # PROOICRS0337
CAS Number:  
Lidocaine
Catalog # PROOICRS0337
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :234.34 g/mol
EINECS:205-302-8
CAS Number:137-58-6
Formula:C₁₄H₂₂N₂O
MDL Number:MFCD00026733