
Benzyltriethylammonium tetrafluoroborate
Catalog # MOLEM73318216
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:279.13 g/mol
Melting Pt:115…116 °C
EINECS:278-768-3
:GHS07
Storage Temperature:Ambient
CAS Number:77794-93-5
Formula:C₁₃H₂₂BF₄N
MDL Number:MFCD00011823