(1R,2S)-(-)-2-Amino-1,2-diphenylethanol
Catalog # MOLE62517904-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:213.28 g/mol
CAS Number:23190-16-1
Formula:C₁₄H₁₅NO
MDL Number:MFCD00074960