Order Entry
Austria
ContactUsLinkComponent
Metanil yellow, Millipore®
Corrosive
Metanil yellow, Millipore®
Catalog # 1.59267.0010
Supplier:  Merck
CAS Number:  
Corrosive
Metanil yellow, Millipore®
Catalog # 1.59267.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    25 g/l (20 °C)
  • Molar Mass:
    375.38 g/mol
  • Bulk Density:
    330 kg/m3
  • pH:
    10 (10 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :375.38 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:209-608-2
 :GHS05
Storage Temperature:Ambient
CAS Number:587-98-4
Formula:C₁₈H₁₄N₃NaO₃S
MDL Number:MFCD00007486