Order Entry
Austria
ContactUsLinkComponent
Pesticide reference standard, 1-Chloronaphthalene 10 µg/ml in isooctane, Dr Ehrenstorfer
Pesticide reference standard, 1-Chloronaphthalene 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20425100IO
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 1-Chloronaphthalene 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20425100IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    90-13-1
  • Description:
    1-Chloronaphthalene 10 µg/ml in isooctane
  • Formula:
    C₁₀H₇Cl
  • Description:
    1-Chloronaphthalene

Specifications

About this item