Farnesol (mixture of isomers)
Catalog # EHERC13405000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:222.37 g/mol
EINECS:225-004-1
CAS Number:4602-84-0
Formula:C₁₅H₂₆O
MDL Number:MFCD00002918