
(R)-(+)-4-Bromo-α-methylbenzylamine 99%, ee 98%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:45791-36-4
:MFCD00066025
Formula:C₈H₁₀BrN
MDL Number:MFCD00025548
:200.08 g/mol
Boiling Pt:140…145 °C (30 mmHg)
Melting Pt:–25 °C
Density:1.371
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:256-245-0
UN:2735
ADR:8,III
:GHS05
Related Information
Frequently Bought Together